missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TREM-2 His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
70.25 € - 217.00 €
Specifications
Accession Number | Q9NZC2 |
---|---|
For Use With (Application) | Control, ELISA, Immunohistochemistry, Western Blot |
Formulation | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gene ID (Entrez) | 54209 |
Name | Human TREM-2 His-tag |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17164253
|
Invitrogen™
A42588 |
50 μg |
217.00 €
50µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17154253
|
Invitrogen™
A42587 |
5 μg |
70.25 €
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).Specifications
Q9NZC2 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human TREM-2 His-tag | |
-20°C | |
<1 EU/μg | |
Trem2 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
54209 | |
HEK293 cells | |
RUO | |
OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; TREM2; TREM-2; Trem2a; Trem2b; Trem2c; triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2 A; triggering receptor expressed on myeloid cells 2 b; triggering receptor expressed on myeloid cells 2 c | |
Human | |
Recombinant | |
≥ 85% |