missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trappin-2/Elafin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF1747-SP
This item is not returnable.
View return policy
Description
Trappin-2/Elafin/Skalp Polyclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Trappin-2/Elafin/Skalp | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cementoin, Elafin, Elastase-specific inhibitor, ESI, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI3, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALP, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, Trappin2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5266 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Neutralization, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 15.6 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| P19957 | |
| PI3 | |
| Mouse myeloma cell line NS0-derived recombinant human Trappin-2/Elafin Ala23-Gln117 Accession # P19957 | |
| 25 μg | |
| Primary | |
| Detects human Trappin-2/Elafin in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) SLPI, rhTGM-2, and human Neutrophil Elastase is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction