missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRA-1-85/CD147 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3195G-100UG
This item is not returnable.
View return policy
Description
TRA-1-85/CD147 Monoclonal specifically detects TRA-1-85/CD147 in Human samples. It is validated for Flow Cytometry.
Specifications
| TRA-1-85/CD147 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| BSG | |
| 2120Ep human embryonal carcinoma cell line | |
| RUO | |
| Detects human TRA-1-85 antigen in Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| TRA-1-85 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction