missing translation for 'onlineSavingsMsg'
Learn More

Human TRA-1-85/CD147 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Product Code. 18770209
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770209 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18770209 Supplier R&D Systems Supplier No. FAB3195U100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRA-1-85/CD147 Monoclonal specifically detects TRA-1-85/CD147 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-85/CD147
Applications Flow Cytometry
Classification Monoclonal
Clone TRA-1-85
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Symbols BSG
Host Species Mouse
Immunogen 2120Ep human embryonal carcinoma cell line
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human TRA-1-85 antigen in Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.