Learn More
Gibco™ Human TNF-RI (soluble) Recombinant Protein
Human TNF-RI (soluble) Recombinant Protein
Brand: Gibco™ PHR3015
Additional Details : Weight : 0.50000kg
Description
Human TNF-RI (soluble) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
sTNF-RI is a member of the TNF-receptor superfamily. This protein is one of the major receptors for the tumor necrosis factor-alpha. This receptor can activate NFκB, mediate apoptosis, and function as a regulator of inflammation. Antiapoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with this receptor, and thus play regulatory roles in the signal transduction mediated by the receptor.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the inhibition of TNF-α mediated cytotoxicity in murine L929 cells in the presence of 0.25ng/mL TNF-α
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
18.3 kDa | |
20 ug | |
Human | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Inflammation, Oncology, Signal Transduction | |
98 % |
7132 | |
Sequential Chromatography | |
Soluble TNF-RI | |
Store in freezer (-5 to -30°C). | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
TNF (Tumor Necrosis Factor) | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.