missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF-alpha Membrane Form PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB210P-025
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry.
Specifications
| TNF-alpha | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7124 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 6401 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P01375 | |
| TNF | |
| E. coli-derived recombinant human TNF-alpha Gly57-Leu233 (predicted) Accession # P01375 | |
| 25 Tests | |
| Primary | |
| Detects cell surface expressed TNF-alpha (membrane and receptor bound forms) by flow cytometry. Detects human TNF-alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant porcine TNF-alpha and recombinant rhesus macaque TNF-alpha is observed but no cross-reactivity with recombinant cotton rat TNF-alpha , recombinant rat TNF-alpha , recombinant human (rh) LT alpha 1/beta 2, rhLT alpha 2/beta 1, rhAPRIL, rhBAFF, rhEDA-A2, recombinant mouse EDA, rhFas Ligand, rhLIGHT, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, or rhVEGI is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction