Learn More
R&D Systems™ Human TMPRSS9 (NP_892018.1) VersaClone cDNA
Specifications
Specifications
| Gene | TMPRSS9 |
| Accession Number | NP_892018.1 |
| Gene ID (Entrez) | 360200 |
| Gene Alias | EC 3.4.21, EC 3.4.21.4, Polyserase 1, polyserase-1, Polyserase-I, Polyserine Protease 1, Transmembrane Protease Serine 9, Transmembrane Protease, Serine 9, Transmembrane Serine Protease 9 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |