missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SYNPO Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
SYNPO Synthetic Peptide, PEP-1176, from Invitrogen.
SYNPO (Synaptopodin) is an actin-associated protein in telencephalic dendrites and renal podocytes. SYNPO is tightly associated with the dendritic spine apparatus, and mice lacking the SYNPO gene lack the apparatus. These SYNPO-null mice also demonstrate an impaired ability to express long-term potentiation as well as deficits in spatial memory tasks, indicating that SYNPO is involved in the regulation of synaptic plasticity. Recent studies suggest that SYNPO is linked to neuronal calcium stores and plays a role in the calcium store-associated ability of neurons to undergo long-term plasticity.
Specifications
Specifications
| Gene ID (Entrez) | 11346 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q8N3V7 |
| Common Name | Human SYNPO |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction