Learn More
Human STRADB (ALS2CR2) knockout HEK-293T cell lysate, 1 Kit
Product Code. 32151993 Shop All Abcam Products
Product Code. 32151993 Supplier Abcam Supplier No. ab2633721Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human STRADB (ALS2CR2) knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: STRADB (ALS2CR2) knockout HEK-293T