Learn More
Abnova™ Human STEAP2 Partial ORF (NP_694544.1, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_694544.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 261729 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172667
|
Abnova™
H00261729-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182667
|
Abnova™
H00261729-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the STEAP family and encodes a multi-pass membrane protein that localizes to the Golgi complex, the plasma membrane, and the vesicular tubular structures in the cytosol. A highly similar protein in mouse has both ferrireductase and cupric reductase activity, and stimulates the cellular uptake of both iron and copper in vitro. Increased transcriptional expression of the human gene is associated with prostate cancer progression. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: ESISMMGSPKSLSETVLPNGINGIKDARKVTVGVIGSGDFAKSLTIRLIRCGYHVVIGSRNPKFASEFFPHVVDVTHHEDALTKTNIIFVAIHREHYTSSpecifications
NP_694544.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IPCA1/PCANAP1/PUMPCn/STAMP1/STMP | |
STEAP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
261729 | |
STEAP2 (Human) Recombinant Protein (Q01) | |
ESISMMGSPKSLSETVLPNGINGIKDARKVTVGVIGSGDFAKSLTIRLIRCGYHVVIGSRNPKFASEFFPHVVDVTHHEDALTKTNIIFVAIHREHYTS | |
RUO | |
STEAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |