missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPACA3 Full-length ORF (NP_776246.1, 1 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00124912-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137643

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Sequence: MVSALRGAPLIRVHSSPVSSPSVSGPRRLVSCLSSQSSALSQSGGGSTSAAGIEARSRALRRRWCPAGIMLLALVCLLSCLLPSSEAKLYGRCELARVLHDFGLDGYRGYSLADWVCLAYFTSGFNAAALDYEADGSTNNGIFQINSRRWCSNLTPNVPNVCRMYCSDLLNPNLKDTVICAMKITQEPQGLGYWEAWRHHCQGKDLTEWVDGCDF
Specifications

Specifications

NP_776246.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
1700025M08Rik/ALLP17/LYC3/LYZL3/MGC119058/SLLP1
SPACA3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
124912
SPACA3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVSALRGAPLIRVHSSPVSSPSVSGPRRLVSCLSSQSSALSQSGGGSTSAAGIEARSRALRRRWCPAGIMLLALVCLLSCLLPSSEAKLYGRCELARVLHDFGLDGYRGYSLADWVCLAYFTSGFNAAALDYEADGSTNNGIFQINSRRWCSNLTPNVPNVCRMYCSDLLNPNLKDTVICAMKITQEPQGLGYWEAWRHHCQGKDLTEWVDGCDF
RUO
SPACA3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions