Learn More
R&D Systems™ Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit
Product Code. 18752404
Change view
Click to view available options
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18752404 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18752404 Supplier R&D Systems™ Supplier No. DSHH00
Please to purchase this item. Need a web account? Register with us today!
Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit quantifies human Sonic Hedgehog/Shh in cell culture supernates (34 ul), serum (34 ul), heparin plasma (34 ul).
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum