missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC19A3 Partial ORF (NP_079519.1, 191 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080704-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100327

  • 335.00 € / 10µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLC19A3 is a member of the reduced folate family of micronutrient transporter genes.[supplied by OMIM]

Sequence: PMPKKSMFFHAKPSREIKKSSSVNPVLEETHEGEAPGCEEQKPTSEILSTSGKLNKGQLNSLKPSNVTVDVFVQWFQDLKECYSSKR
Specifications

Specifications

NP_079519.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.31kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PMPKKSMFFHAKPSREIKKSSSVNPVLEETHEGEAPGCEEQKPTSEILSTSGKLNKGQLNSLKPSNVTVDVFVQWFQDLKECYSSKR
RUO
SLC19A3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
80704
SLC19A3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
THTR2
SLC19A3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions