missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 LlaMABodyTM Bivalent VHH Llama IgG2 Fusion Antibody, R&D Systems™

Llama Monoclonal Antibody
Brand: R&D Systems LMAB10902-100
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
| Mouse myeloma cell line NS0-derived human Siglec-3/CD33 protein as immunogen for bivalent Llama VHH-Llama IgG2 Fusion Antibody, Asp18-His259, Accession # AAA51948 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Flow Cytometry | |
| L008.2.25LL | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| AAA51948 | |
| Llama | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 945 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction