missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SDHA Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
234.00 € - 536.00 €
Specifications
| Antigen | SDHA |
|---|---|
| Clone | 1084056 |
| Dilution | Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30241047
|
R&D Systems
MAB11586-100 |
100 μg |
536.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30241005
|
R&D Systems
MAB11586-SP |
25 μg |
234.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SDHA Monoclonal antibody specifically detects SDHA in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,ImmunocytochemistrySpecifications
| SDHA | |
| Western Blot 1 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| CMD1GG, flavoprotein subunit of complex II, FP, MC2DN1, NDAXOA, PGL5, PPGL5, SDH1, SDH2, SDHF, succinate dehydrogenase complex, subunit A, flavoprotein (Fp) | |
| Synthetic Peptide, Accession # P31040 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1084056 | |
| Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P31040 | |
| 6389 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title