missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SCF/c-kit Ligand Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF-255-SP
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| SCF/c-kit Ligand | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 4254 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.05-0.3 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| P21583 | |
| KITLG | |
| E. coli-derived recombinant human SCF/c-kit Ligand, Glu26-Ala189, Accession # P21583 | |
| 25 μg | |
| Primary | |
| Detects human SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs, less than 60% cross-reactivity with recombinant mouse SCF is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction