missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SASS6 Full-length ORF (AAI01027.1, 1 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAI01027.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 163786 |
Molecular Weight (g/mol) | 81.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148983
|
Abnova™
H00163786-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138983
|
Abnova™
H00163786-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SAS6 is necessary for centrosome duplication and functions during procentriole formation; SAS6 functions to ensure that each centriole seeds the formation of a single procentriole per cell cycle Strnad et al., (2007) [PubMed 17681132].[supplied by OMIM]
Sequence: MQSLDDATKQLDFTRKTLAEKKQELDKLRNEWASHTAALTNKHSQELTNEKEKALQAQVQYQQQHEQQKKDLEILHQQNIHQLQNRLSELEVANKDLTERKYKGDSTIRELKAKLSGVEEELQRTKQEVLSLRRENSTLDVECHEKEKHVNQLQTKVAVLEQEIKDKDQLVLRTKEAFDTIQEQKVVLEENGEKNQVQLGKLEATIKSLSAELLKANEIIKKLQGDLKTLMGKLKLKNTVTIQQEKLLAEKEEKLQKEQKELQDVGQSLRIKEQEVCKLQEQLEATVKKLEESKQLLKNNEKLITWLNKELNENQLVRKQDVLGPSTTPPAHSSSNTIRSGISPNLNVVDGRLTYPTCGIGYPVSSAFAFQNTFPHSISAKNTSHPGSGTKVQFNLQFTKPNASLGDVQSGATISMPCSTDKENGENVGLESKYLKKREDSIPLRGLSQNLFSNSDHQRDGTLGALHTSSKPTALPSASSAYFPGQLPNSSpecifications
AAI01027.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQSLDDATKQLDFTRKTLAEKKQELDKLRNEWASHTAALTNKHSQELTNEKEKALQAQVQYQQQHEQQKKDLEILHQQNIHQLQNRLSELEVANKDLTERKYKGDSTIRELKAKLSGVEEELQRTKQEVLSLRRENSTLDVECHEKEKHVNQLQTKVAVLEQEIKDKDQLVLRTKEAFDTIQEQKVVLEENGEKNQVQLGKLEATIKSLSAELLKANEIIKKLQGDLKTLMGKLKLKNTVTIQQEKLLAEKEEKLQKEQKELQDVGQSLRIKEQEVCKLQEQLEATVKKLEESKQLLKNNEKLITWLNKELNENQLVRKQDVLGPSTTPPAHSSSNTIRSGISPNLNVVDGRLTYPTCGIGYPVSSAFAFQNTFPHSISAKNTSHPGSGTKVQFNLQFTKPNASLGDVQSGATISMPCSTDKENGENVGLESKYLKKREDSIPLRGLSQNLFSNSDHQRDGTLGALHTSSKPTALPSASSAYFPGQLPNS | |
RUO | |
SASS6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
163786 | |
SASS6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22097/MGC119440/SAS-6/SAS6 | |
SASS6 | |
Recombinant | |
wheat germ expression system |