missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SAGE1 Partial ORF (NP_061136, 357 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055511-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138176

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas. [provided by RefSeq]

Sequence: PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN
Specifications

Specifications

NP_061136
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN
RUO
SAGE1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55511
SAGE1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SAGE
SAGE1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions