missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human S100A8/S100A9 Heterodimer DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human S100A8/S100A9 Heterodimer DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human S100A8/S100A9 Heterodimer.
Specifications
Specifications
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Binding Target | S100A8/S100A9 Heterodimer |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction