missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RPS6KA5 Partial ORF (AAH17187.1, 372 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009252-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: FQGYSFVAPSILFKRNAAVIDPLQFHMGVERPGVTNVARSAMMKDSPFYQHYDLDLKDKPLGEGSFSICRKCVHKKSNQAFAVKIISKRMEANTQKEITSpecifications
AAH17187.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FQGYSFVAPSILFKRNAAVIDPLQFHMGVERPGVTNVARSAMMKDSPFYQHYDLDLKDKPLGEGSFSICRKCVHKKSNQAFAVKIISKRMEANTQKEIT | |
RUO | |
RPS6KA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9252 | |
RPS6KA5 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC1911/MSK1/MSPK1/RLPK | |
RPS6KA5 | |
Recombinant | |
wheat germ expression system |