missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF103 Full-length ORF (AAH35053, 1 a.a. - 685 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH35053 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7844 |
Molecular Weight (g/mol) | 101.09kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16192142
|
Abnova™
H00007844-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182142
|
Abnova™
H00007844-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene contains a RING-H2 finger, a motif known to be involved in protein-protein and protein-DNA interactions. This gene is highly expressed in normal cerebellum, but not in the cerebral cortex. The expression of the rat counterpart in the frontal cortex and hippocampus was shown to be induced by elctroconvulsive treatment (ECT) as well as chronic antidepressant treatment, suggesting that this gene may be a molecular target for ECT and antidepressants. [provided by RefSeq]
Sequence: MWLKLFFLLLYFLVLFVLARFFEAIVWYETGIFATQLVDPVALSFKKLKTILECRGLGYSGLPEKKDVRELVEKSGDLMEGELYSALKEEEASESVSSTNFSGEMHFYELVEDAKDGIWLVQVIANDRSPLVGKIHWEKMVKKVSRFGIRTGTFNCSSDPRYCRRRGWVRSTLIMSVPQTSTSKGKVMLKEYSGRKIEVEHIFKWITAHAASRIKTIYNAEHLKEEWNKSDQYWLKIYLFANLDQPPAFFPALSIKFTGRVEFIFVNVENWDNKSYMTDIGIYNMPSYILRTPEGIYRYGNHTGEFISLQAMDSFLRSLQPEVNDLFVLSLVLVNLMAWMDLFITQGATIKRFVVLISTLGTYNSLLIISWLPVLGFLQLPYLDSFYEYSLKLLRYSNTTTLASWVRADWMFYSSHPALFLSTYLGHGLLIDYFEKKRRRNNNNDEVNANNLEWLSSLWDWYTSYLFHPIASFQNFPVESDWDEDPDLFLERLAFPDLWLHHLIPTDYIKNLPMWRFKCLGVQSEEEMSEGSQDTENDSESENTDTLSSEKEVFEDKQSVLHNSPGTASHCDAEACSCANKYCQTSPCERKGRSYGSYNTNEDMEPDWLTWPADMLHCTECVVCLENFENGCLLMGLPCGHVFHQNCIVMWLAGGRHCCPVCRWPSYKKKQPYAQHQPLSNDVPSSpecifications
AAH35053 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.09kDa | |
Glutathione Sepharose 4 Fast Flow | |
MWLKLFFLLLYFLVLFVLARFFEAIVWYETGIFATQLVDPVALSFKKLKTILECRGLGYSGLPEKKDVRELVEKSGDLMEGELYSALKEEEASESVSSTNFSGEMHFYELVEDAKDGIWLVQVIANDRSPLVGKIHWEKMVKKVSRFGIRTGTFNCSSDPRYCRRRGWVRSTLIMSVPQTSTSKGKVMLKEYSGRKIEVEHIFKWITAHAASRIKTIYNAEHLKEEWNKSDQYWLKIYLFANLDQPPAFFPALSIKFTGRVEFIFVNVENWDNKSYMTDIGIYNMPSYILRTPEGIYRYGNHTGEFISLQAMDSFLRSLQPEVNDLFVLSLVLVNLMAWMDLFITQGATIKRFVVLISTLGTYNSLLIISWLPVLGFLQLPYLDSFYEYSLKLLRYSNTTTLASWVRADWMFYSSHPALFLSTYLGHGLLIDYFEKKRRRNNNNDEVNANNLEWLSSLWDWYTSYLFHPIASFQNFPVESDWDEDPDLFLERLAFPDLWLHHLIPTDYIKNLPMWRFKCLGVQSEEEMSEGSQDTENDSESENTDTLSSEKEVFEDKQSVLHNSPGTASHCDAEACSCANKYCQTSPCERKGRSYGSYNTNEDMEPDWLTWPADMLHCTECVVCLENFENGCLLMGLPCGHVFHQNCIVMWLAGGRHCCPVCRWPSYKKKQPYAQHQPLSNDVPS | |
RUO | |
RNF103 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7844 | |
RNF103 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KF1/MGC102815/MGC41857/ZFP103/hkf-1 | |
RNF103 | |
Recombinant | |
wheat germ expression system |