missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant LIF
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- Leukemia inhibitory factor (LIF) is an interleukin 6 class cytokine that regulates a broad variety of developmental functions
- After LIF binds to LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling
- LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype
- Produced by the endometrium, LIF plays an important autocrine and paracrine role in implantation by regulating proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment
- Human LIF can be used for the maintenance of mouse embryonic stem cells, however mouse LIF cannot bind to the human receptor, thus rendering mouse LIF inactive
- LIF is produced by CD4+ and activated regulatory T cells, and promotes Foxp3 expression, while repressing Th17 lineage-specific genes
Specifications
Specifications
| Name | CDF, Cholinergic differentiation factor, D Factor, DIA, Differentiation-inducing factor, Differentiation inhibitory activity, Differentiation-stimulating factor, Emfilermin, Hepatocyte-stimulating factor III, HILDA, Leukemia inhibitory factor, MLPLI |
| Quantity | 50 μg |
| Cell Type | Neural Cells, PSC-Derived; Neurons; Pluripotent Stem Cells |
| Purity or Quality Grade | >95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction