missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-2, ACF
Description
- Interleukin 2 (IL-2) is a monomeric cytokine that was originally identified as a T cell growth factor
- It binds to heterotrimeric receptors consisting of CD25, CD122, and CD132
- Upon binding, it activates JAK3-, STAT5-, and AKT-dependent signaling pathways, which results in cellular proliferation and survival (Ma et al.) The majority of IL-2 is secreted by activated CD4+ and CD8+ T cells, although B cells and dendritic cells were found to produce IL-2 in small amounts
- IL-2 downregulates immune responses to prevent autoimmunity during thymic development, influences the development of CD4+CD25+ T regulatory cells and affects development of follicular helper T cells
- IL-2 also controls inflammation by inhibiting Th17 differentiation
- This product is animal component-free
Specifications
Specifications
| Name | Aldesleukin, Interleukin-2, T cell growth factor, TCGF |
| Quantity | 10 μg |
| Cell Type | Hematopoietic Stem and Progenitor Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+ |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?