Learn More
STEMCELL Technologies Human Recombinant EGFR
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
- Epidermal growth factor receptor (EGFR) is a type I transmembrane protein and receptor tyrosine kinase
- EGFR has been shown to bind to some members of the EGF family ligands including EGF, amphiregulin, TGF-α, betacellulin, epiregulin, heparin-binding EGF, and neuregulin-2α
- EGFR ligand binding induces homodimerization, as well as heterodimerization of EGFR with ErbB2 or with ligand-activated ErbB3 or ErbB4 (Schlessinger)
- Dimerization results in kinase activation, phosphorylation, and cell signaling, mediated primarily through MEK/ERF and AKT pathways (Navlonic et al.) EGFR signaling has been shown to regulate cell proliferation, differentiation, motility, and apoptosis
- Elevated levels of EGFR have been correlated with carcinogenesis (Maihle et al.) Protein contains a His-residue tag at the carboxyl end of the polypeptide chain
Specifications
Specifications
| Name | Avian erythroblastic leukemia viral oncogene homolog, Epidermal growth factor receptor, Erb-b2 receptor tyrosine kinase 1, ERBB1, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1, v-erb-b |
| Quantity | 50 μg |
| Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |