missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant BMP-4

Product Code. 17109851
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
20 μg
Unit Size:
1000µg
100µg
20µg
This item is not returnable. View return policy

Product Code. 17109851

Brand: STEMCELL Technologies 78211.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bone morphogenetic protein 4

  • Bone morphogenetic protein 4 (BMP-4) is a member of the highly conserved transforming growth factor β (TGF-β) superfamily
  • Mature BMP-4 is a disulfide-linked homodimeric protein consisting of two 116-amino-acid residue subunits, and is generated by the proteolytic removal of the signal peptide and propeptides (Xiao et al.) BMP-4 binds to type I and type II receptors on cells
  • This binding results in the phosphorylation of receptor 1, which in turn results in the phosphorylation of Smad proteins, which then go on to act as transcription factors (Zhang et al.) BMPs have been shown to be key regulators of embryogenesis and are known to play a role in the growth and differentiation of various cell types, including embryonic stem (ES) cells, induced pluripotent stem (iPS) cells, mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells (Chadwick et al.; Graham et al.; Jones et al.; Lengerke et al.; Zhang et al.)

Specifications

Name BMP-2B, BMP2B1, MCOPS6, OFC11, ZYME
Quantity 100 μg
Cell Type Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.