missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human RCBTB2 - Ready-To-Use ELISA Kit (Colorimetric)

Product Code. 30239033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30239033 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30239033 Supplier Novus Biologicals™ Supplier No. NBP340325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.312 - 20 ng/mL (example only; lot dependent)
Assay Sensitivity 0.124 ng/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 1102
Gene Symbol RCBTB2
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CHC1-L, CHC1LRCC1-like G exchanging factor RLG, Chromosome condensation 1-like, RCC1 and BTB domain-containing protein 2, RCC1-like G exchanging factor, regulator of chromosome condensation (RCC1) and BTB (POZ) domain containingprotein 2, regulator of chromosome condensation and BTB domain containing protein 2, Regulator of chromosome condensation and BTB domain-containing protein 2, RLG
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.