missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Fibronectin Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF1918
This item is not returnable.
View return policy
Description
Fibronectin Polyclonal specifically detects Fibronectin in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| Fibronectin | |
| Polyclonal | |
| Unconjugated | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2335 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| FN1 | |
| Human plasma-derived Fibronectin | |
| 100 μg | |
| Primary | |
| Detects human Fibronectin in direct ELISAs. Detects human and rat Fibronectin in Western blots. | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction