missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RASIP1 Full-length ORF (AAH21860.1, 1 a.a. - 317 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number AAH21860.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54922
Molecular Weight (g/mol) 61.7kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16100903
View Documents Promotion Details
Abnova™
H00054922-P01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16110903
View Documents Promotion Details
Abnova™
H00054922-P01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MLWMANTTELLSFVQEKVLEMEKEADQEDPQLCNDLELCDEAMALLDEVIMCTFQQSVYYLTKTLYSTLPALLDSNPFTAGAELPGPGAELGAMPPGLRPTLGVFQAALELTSQCELHPDLVSQTFGYLFFFSNASLLNSLMERGQGRPFYQWSRAVQIRTNLDLVLDWLQGAGLGDIATEFFRKLSMAVNLLCVPRTSLLKASWSSLRTDHPTLTPAQLHHLLSHYQLGPGRGPPAAWDPPPAEREAVDTGDIFESFSSHPPLILPLGSSRLRLTGPVTDDALHRELRRLRRLLWDLEQQELPANYRHGPPVATSP
Specifications

Specifications

AAH21860.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.7kDa
Glutathione Sepharose 4 Fast Flow
MLWMANTTELLSFVQEKVLEMEKEADQEDPQLCNDLELCDEAMALLDEVIMCTFQQSVYYLTKTLYSTLPALLDSNPFTAGAELPGPGAELGAMPPGLRPTLGVFQAALELTSQCELHPDLVSQTFGYLFFFSNASLLNSLMERGQGRPFYQWSRAVQIRTNLDLVLDWLQGAGLGDIATEFFRKLSMAVNLLCVPRTSLLKASWSSLRTDHPTLTPAQLHHLLSHYQLGPGRGPPAAWDPPPAEREAVDTGDIFESFSSHPPLILPLGSSRLRLTGPVTDDALHRELRRLRRLLWDLEQQELPANYRHGPPVATSP
RUO
RASIP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
54922
RASIP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20401/RAIN
RASIP1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications