missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RASGRP1 (aa 696-795) Control Fragment Recombinant Protein

Product Code. 30201466
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201466 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30201466 Supplier Invitrogen™ Supplier No. RP106113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83459 (PA5-83459. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Functions as a calcium- and diacylglycerol (DAG)-regulated nucleotide exchange factor specifically activating Ras through the exchange of bound GDP for GTP. Activates the Erk/MAP kinase cascade. Regulates T-cell/B-cell development, homeostasis and differentiation by coupling T-lymphocyte/B-lymphocyte antigen receptors to Ras. Regulates NK cell cytotoxicity and ITAM-dependent cytokine production by activation of Ras-mediated ERK and JNK pathways. Functions in mast cell degranulation and cytokine secretion, regulating FcERI-evoked allergic responses. May also function in differentiation of other cell types.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95267
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10125
Name Human RASGRP1 (aa 696-795) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor II; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor II; CALDAG-GEFI; CALDAG-GEFII; guanine nucleotide exchange factor, calcium- and DAG-regulated, Rap1A; hRasGRP1; ras activator RasGRP; RAS guanyl nucleotide-releasing protein 1; RAS guanyl releasing protein 1; RAS guanyl releasing protein 1 (calcium and DAG-regulated); Ras guanyl-releasing protein; RAS guanyl-releasing protein 1; RASGRP; RASGRP1; V
Common Name RASGRP1
Gene Symbol RASGRP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RKTAQDTLYVLPSPTSPCPSPVLVRKRAFVKWENKDSLIKSKEELRHLRLPTYQELEQEINTLKADNDALKIQLKYAQKKIESLQLEKSNHVLAQMEQGD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.