Learn More
Human RAB14 knockout HEK-293T cell lysate, 1 Kit
Product Code. 32141184 Shop All Abcam Products
Product Code. 32141184 Supplier Abcam Supplier No. ab2581601Kit
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human RAB14 knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: RAB14 knockout HEK-293T