missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PSTPIP1 Partial ORF (NP_003969, 148 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003969 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9051 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139815
|
Abnova™
H00009051-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129815
|
Abnova™
H00009051-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene binds to the cytoplasmic tail of CD2, an effector of T cell activation and adhesion, negatively affecting CD2-triggered T cell activation. The encoded protein appears to be a scaffold protein and a regulator of the actin cytoskeleton. It has also been shown to bind ABL1, PTPN18, WAS, CD2AP, and PTPN12. Mutations in this gene are a cause of PAPA syndrome. [provided by RefSeq]
Sequence: CRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDESpecifications
NP_003969 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD2BP1/CD2BP1L/CD2BP1S/H-PIP/PAPAS/PSTPIP | |
PSTPIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9051 | |
PSTPIP1 (Human) Recombinant Protein (Q01) | |
CRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDE | |
RUO | |
PSTPIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |