missing translation for 'onlineSavingsMsg'
Learn More

Human Prostaglandin I2 Synthase Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18719297
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18719297 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18719297 Supplier R&D Systems Supplier No. AF7788SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Prostaglandin I2 Synthase Polyclonal specifically detects Prostaglandin I2 Synthase in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostaglandin I2 Synthase
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16647
Gene Alias CYP8, CYP8A1, CYP8A1PTGI, CYP8prostacyclin synthase, EC 5.3.99.4, MGC126858, MGC126860, PGIS, PGIScytochrome P450, family 8, subfamily A, polypeptide 1, Prostacyclin Synthase, prostaglandin I2 (prostacyclin) synthase, Prostaglandin I2 synthase, PTGI
Gene Symbols PTGIS
Host Species Sheep
Immunogen E. coli-derived recombinant human Prostaglandin I2 Synthase Arg21-Pro500 Accession # Q16647
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5740
Test Specificity Detects human Prostaglandin I2 Synthase in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.