missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3175
This item is not returnable.
View return policy
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Immunohistochemistry.
Specifications
| BDNF | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P23560 | |
| BDNF | |
| Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Ala19-Arg247 Accession # P23560 | |
| 100 μg | |
| Primary | |
| Detects human Pro-BDNF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BDNF (mature), rh beta-NGF, rhNT-3, rhNT-4, rhPro-NT-3 or rhPro-NT-4 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 584421 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 627 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction