missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PMS1 Partial ORF (NP_000525, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005378-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16129251

  • 335.00 € / 10µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome. [provided by RefSeq]

Sequence: KELIENSLDAGATSVDVKLENYGFDKIEVRDNGEGIKAVDAPVMAMKYYTSKINSHEDLENLTTYGFRGEALGSICCIAEVLITTRTAADNFSTQYVLDGSGHILSQKPS
Specifications

Specifications

NP_000525
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KELIENSLDAGATSVDVKLENYGFDKIEVRDNGEGIKAVDAPVMAMKYYTSKINSHEDLENLTTYGFRGEALGSICCIAEVLITTRTAADNFSTQYVLDGSGHILSQKPS
RUO
PMS1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5378
PMS1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781M0253/FLJ98259/HNPCC3/PMSL1/hPMS1
PMS1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions