missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Pluripotent Stem Cell Naïve State qPCR Array

Product Code. p-7217566
Encompass_Preferred
Click to view available options
Format:
96-well Plate Type 3
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17129291

Brand: STEMCELL Technologies 07523

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For characterization of gene expression associated with hPSC naïve or primed states

  • Characterize your hPSCs by accurately measuring their gene expression profile to determine their status along the spectrum from naïve to primed pluripotency
  • The Human Pluripotent Stem Cell (hPSC) Naïve State qPCR Array has validated primers and probes to detect 90 genes preselected for their demonstrated differential expression in naïve state and primed hPSCs and their derivatives, as well as 6 endogenous control genes
  • TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control
  • Naïve state hPSCs are self-renewing, retain the properties of the pre-implantation blastocyst, and can differentiate to both embryonic and extra-embryonic lineages
  • Primed hPSCs are also self-renewing, but they are restricted to differentiation along the three embryonic germ layers

Specifications

Description Pluripotent Stem Cells
Format 96-well Plate Type 3
For Use With (Application) Real Time PCR (qPCR)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.