missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PLA2G7/PAF-AH/Lp-PLA2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5106-SP
This item is not returnable.
View return policy
Description
PLA2G7/PAF-AH/Lp-PLA2 Polyclonal specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| PLA2G7/PAF-AH/Lp-PLA2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LpPLA2, LP-PLA2, PAF acetylhydrolase, PAF-AH, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7941 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAH38452 | |
| PLA2G7 | |
| E. coli-derived recombinant human PLA2G7/PAF-AH/Lp-PLA2 Met33-Asn441 Accession # AAH38452 | |
| 25 μg | |
| Primary | |
| Detects human PLA2G7/PAF-AH/Lp-PLA2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) PLA2G-2A, recombinant mouse PLA2G-2A, and rhPLA2G-1B is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction