missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLA2G4A (aa 656-746) Control Fragment Recombinant Protein

Product Code. 30200692
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30200692 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30200692 Supplier Invitrogen™ Supplier No. RP103178

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111389 (PA5-111389. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the cytosolic phospholipase A2 group IV family. The enzyme catalyzes the hydrolysis of membrane phospholipids to release arachidonic acid which is subsequently metabolized into eicosanoids. Eicosanoids, including prostaglandins and leukotrienes, are lipid-based cellular hormones that regulate hemodynamics, inflammatory responses, and other intracellular pathways. The hydrolysis reaction also produces lysophospholipids that are converted into platelet-activating factor. The enzyme is activated by increased intracellular Ca(2+) levels and phosphorylation, resulting in its translocation from the cytosol and nucleus to perinuclear membrane vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P47712
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5321
Name Human PLA2G4A (aa 656-746) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias calcium-dependent phospholipid-binding protein; Cpla2; cPLA2alpha; cPLA2-alpha; cytosolic group IVA; Cytosolic phospholipase A2; group IVA; lysophospholipase; MGC126350; Phosphatidylcholine 2-acylhydrolase; Phospholipase A2; phospholipase A2 cytosolic; phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent); Pla2c; Pla2g4; PLA2G4A
Common Name PLA2G4A
Gene Symbol PLA2G4A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PRETEEEKEIADFDIFDDPESPFSTFNFQYPNQAFKRLHDLMHFNTLNNIDVIKEAMVESIEYRRQNPSRCSVSLSNVEARRFFNKEFLSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.