missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Chk2 (T68) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF1626-SP
This item is not returnable.
View return policy
Description
Chk2 Polyclonal specifically detects Chk2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Chk2 (p thr68) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHEK2 | |
| Phosphopeptide containing human Chk2 T68 site | |
| 25 μg | |
| Primary | |
| Detects human Chk2 when phosphorylated at T68. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11200 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction