missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PGGT1B Partial ORF (NP_005014, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005229-Q01S

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172695

  • 335.00 € / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MVATEDERLAGSGEGERLDFLRDRHVRFFQRCLQVLPERYSSLETSRLTIAFFALSGLDMLDSLDVVNKDDIIEWIYSLQVLPTEDRSNLNRCGFRGSSYLGIPF
Specifications

Specifications

NP_005014
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVATEDERLAGSGEGERLDFLRDRHVRFFQRCLQVLPERYSSLETSRLTIAFFALSGLDMLDSLDVVNKDDIIEWIYSLQVLPTEDRSNLNRCGFRGSSYLGIPF
RUO
PGGT1B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5229
PGGT1B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BGGI/GGTI
PGGT1B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions