missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Pericentrin (aa 373-517) Control Fragment Recombinant Protein

Product Code. 30195717
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195717

Brand: Invitrogen™ RP91790

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53498 (PA5-53498. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95613
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5116
Name Human Pericentrin (aa 373-517) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW476095; C86676; KEN; kendrin; KIAA0402; m239Asp; m275Asp; MOPD2; PCN; Pcnt; PCNT2; PCNTB; PCTN2; Pericentrin; pericentrin (kendrin); pericentrin 2; pericentrin-2; pericentrin-250; pericentrin-360; pericentrin-380; pericentrin-B; SCKL4
Common Name Pericentrin
Gene Symbol PCNT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEDLQNQFQKELAEQRAELEKIFQDKNQAERALRNLESHHQAAIEKLREDLQSEHGRCLEDLEFKFKESEKEKQLELENLQASYEDLKAQSQEEIRRLWSQLDSARTSRQELSELHEQLLARTSRVEDLEQLKQREKTQHESELE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.