missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCOLN3 Full-length ORF (-, 1 a.a. - 196 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005119-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138801

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

This gene encodes a member of the CHMP/Chmp family of proteins which are involved in multivesicular body sorting of proteins to the interiors of lysosomes. The initial prediction of the protein sequence encoded by this gene suggested that the encoded protein was a metallopeptidase. The nomenclature has been updated recently to reflect the correct biological function of this encoded protein. [provided by RefSeq]

Sequence: MDDTLFQLKFTAKQLEKLAKKAEKDSKAEQAKVKKALLQKNVECARVYAENAIRKKNEGVNWLRMASRVDAVASKVQTAVTMKGVTKNMAQVTKALDKALSTMDLQKVSSVMDRFEQQVQNLDVHTSVMEDSMSSATTLTTPQEQVDSLIMQIAEENGLEVLDQLSQLPEGASAVGESSVRSQEDQLSRRLAALRN
Specifications

Specifications

Antibody Production, Protein Array, ELISA, Western Blot
5119
PCOLN3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDTLFQLKFTAKQLEKLAKKAEKDSKAEQAKVKKALLQKNVECARVYAENAIRKKNEGVNWLRMASRVDAVASKVQTAVTMKGVTKNMAQVTKALDKALSTMDLQKVSSVMDRFEQQVQNLDVHTSVMEDSMSSATTLTTPQEQVDSLIMQIAEENGLEVLDQLSQLPEGASAVGESSVRSQEDQLSRRLAALRN
RUO
CHMP1A
Wheat Germ (in vitro)
GST
Liquid
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.19kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CHMP1/KIAA0047/PCOLN3/PRSM1
CHMP1A
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions