missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PARVA Full-length ORF (NP_060692.1, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055742-P02.10ug
Additional Details : Weight : 0.00010kg
Description
Members of the parvin family, including PARVA, are actin-binding proteins associated with focal contacts.[supplied by OMIM]
Sequence: MATSPQKSPSVPKSPTPKSPPSRKKDDSFLGKLGGTLARRKKAKEVSELQEEGMNAINLPLSPIPFELDPEDTMLEENEVRTMVDPNSRSDPKLQELMKVLIDWINDVLVGERIIVKDLAEDLYDGQVLQKLFEKLESEKLNVAEVTQSEIAQKQKLQTVLEKINETLKLPPRSIKWNVDSVHAKSLVAILHLLVALSQYFRAPIRLPDHVSIQVVVVQKREGILQSRQIQEEITGNTEALSGRHERDAFDTLFDHAPDKLNVVKKTLITFVNKHLNKLNLEVTELETQFADGVYLVLLMGLLEGYFVPLHSFFLTPDSFEQKVLNVSFAFELMQDGGLEKPKPRPEDIVNCDLKSTLRVLYNLFTKYRNVESpecifications
NP_060692.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10793/FLJ12254/MXRA2 | |
PARVA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55742 | |
PARVA (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATSPQKSPSVPKSPTPKSPPSRKKDDSFLGKLGGTLARRKKAKEVSELQEEGMNAINLPLSPIPFELDPEDTMLEENEVRTMVDPNSRSDPKLQELMKVLIDWINDVLVGERIIVKDLAEDLYDGQVLQKLFEKLESEKLNVAEVTQSEIAQKQKLQTVLEKINETLKLPPRSIKWNVDSVHAKSLVAILHLLVALSQYFRAPIRLPDHVSIQVVVVQKREGILQSRQIQEEITGNTEALSGRHERDAFDTLFDHAPDKLNVVKKTLITFVNKHLNKLNLEVTELETQFADGVYLVLLMGLLEGYFVPLHSFFLTPDSFEQKVLNVSFAFELMQDGGLEKPKPRPEDIVNCDLKSTLRVLYNLFTKYRNVE | |
RUO | |
PARVA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |