Learn More
Human PARP10 (ARTD10) knockout A549 cell lysate, 1 Kit
Product Code. 32143438 Shop All Abcam Products
Product Code. 32143438 Supplier Abcam Supplier No. ab2585631Kit
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human PARP10 (ARTD10) knockout A549 cell/tissue lysate.
  • Species: Human
  • Tissue: PARP10 (ARTD10) knockout A549