missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PANX1 Full-length ORF (AAH16931.1, 1 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00024145-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137712

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

The protein encoded by this gene belongs to the innexin family. Innexin family members are the structural components of gap junctions. This protein and pannexin 2 are abundantly expressed in central nerve system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that this protein alone and in combination with pannexin 2 may form cell type-specific gap junctions with distinct properties. [provided by RefSeq]

Sequence: MAIAHLATEYVFSDFLLKEPTEPKFKGLRLELAVDKMVTCIAVGLPLLLISLAFAQEISIGTQISCFSPSSFSWRQAAFVDSYCWAAVQQKNSLQSESGNLPLWLHKFFPYILLLFAILLYLPPLFWRFAAAPHICSDLKFIMEELDKVYNRAIKAAKSARDLDMRDGACSVPGVTENLGQSLWEVSESHFKYPIVEQYLKTKKNSNNLIIKYISCRLLTLIIILLACIYLGYYFSLSSLSDEFVCSIKSGILRNDSTVPDQFQCKLIAVGIFQLLSVINLVVYVLLAPVVVYTLFVPFRQKTDVLKVYEILPTFDVLHFKSEGYNDLSLYNLFLEENISEVKSYKCLKVLENIKSSGQGIDPMLLLTNLGMIKMDVVDGKTPMSAEMREEQGNQTAELQGMNIDSETKANNGEKNARQRLLDSSC
Specifications

Specifications

AAH16931.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC21309/MRS1/PX1/UNQ2529
PANX1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
24145
PANX1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAIAHLATEYVFSDFLLKEPTEPKFKGLRLELAVDKMVTCIAVGLPLLLISLAFAQEISIGTQISCFSPSSFSWRQAAFVDSYCWAAVQQKNSLQSESGNLPLWLHKFFPYILLLFAILLYLPPLFWRFAAAPHICSDLKFIMEELDKVYNRAIKAAKSARDLDMRDGACSVPGVTENLGQSLWEVSESHFKYPIVEQYLKTKKNSNNLIIKYISCRLLTLIIILLACIYLGYYFSLSSLSDEFVCSIKSGILRNDSTVPDQFQCKLIAVGIFQLLSVINLVVYVLLAPVVVYTLFVPFRQKTDVLKVYEILPTFDVLHFKSEGYNDLSLYNLFLEENISEVKSYKCLKVLENIKSSGQGIDPMLLLTNLGMIKMDVVDGKTPMSAEMREEQGNQTAELQGMNIDSETKANNGEKNARQRLLDSSC
RUO
PANX1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions