Learn More
Human Olfactomedin-4/OLFM4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9218-SP
This item is not returnable.
View return policy
Description
OLFM4 Monoclonal specifically detects OLFM4 in Human samples. It is validated for Western Blot.
Specifications
| OLFM4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLFM4, OLM4, UNQ362 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Olfactomedin-4/OLFM4 Asp21-Gln510 Accession # Q6UX06 | |
| 25 μg | |
| Primary | |
| Detects human Olfactomedin-4/OLFM4 in direct ELISAs and Western blots. | |
| Human | |
| IgG1 |
| Western Blot | |
| 806305 | |
| Western Blot 2 ug/mL | |
| Q6UX06 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10562 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |