missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NPY Full-length ORF (AAH29497, 1 a.a. - 97 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004852-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138431

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Sequence: MLGNKRLGLSGLTLALSLLVCLGALAEAYPSKPDNPGEDAPAEDMARYYSALRHYINLITRQRYGKRSSPETLISDLLMRESTENVPRTRLEDPAMW
Specifications

Specifications

AAH29497
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PYY4
NPY
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4852
NPY (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLGNKRLGLSGLTLALSLLVCLGALAEAYPSKPDNPGEDAPAEDMARYYSALRHYINLITRQRYGKRSSPETLISDLLMRESTENVPRTRLEDPAMW
RUO
NPY
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions