missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2C/CD159c Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB138N-100
This item is not returnable.
View return policy
Description
NKG2C/CD159c/KLRC2 Monoclonal specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKG2C/CD159c/KLRC2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| KLRC2 | |
| BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94 | |
| 100 Tests | |
| Primary | |
| Detects human NKG2C/CD159c heterodimer with CD94 in flow cytometry. It does not cross-react with the human NKG2A/CD94 heterodimer or with the human CD94 homodimer. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 134591 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, KLRC2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3822 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction