missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Nidogen Recombinant Protein, PeproTech®

Product Code. 30019830
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019830 25 μg 25µg
30019789 5 μg 5µg
30019358 500 μg 500µg
30019712 250 μg 250µg
30019982 1 mg 1mg
30019473 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30019830 Supplier Gibco™ Supplier No. 1401725UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14543
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 4811
Molecular Weight (g/mol) 133.5 kDa
Name Human Nidogen
Quantity 25 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Common Name Nidogen
Biological Activity Determined by the ability of the immobilized protein to support the adhesion of HUVEC cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence LSRQELFPFG PGQGDLELED GDDFVSPALE LSGALRFYDR SDIDAVYVTT NGIIATSEPP AKESHPGLFP PTFGAVAPFL ADLDTTDGLG KVYYREDLSP SITQRAAECV HRGFPEISFQ PSSAVVVTWE SVAPYQGPSR DPDQKGKRNT FQAVLASSDS SSYAIFLYPE DGLQFHTTFS KKENNQVPAV VAFSQGSVGF LWKSNGAYNI FANDRESVEN LAKSSNSGQQ GVWVFEIGSP ATTNGVVPAD VILGTEDGAE YDDEDEDYDL ATTRLGLEDV GTTPFSYKAL RRGGADTYSV PSVLSPRRAA TERPLGPPTE RTRSFQLAVE TFHQQHPQVI DVDEVEETGV VFSYNTDSRQ TCANNRHQCS VHAECRDYAT GFCCSCVAGY TGNGRQCVAE GSPQRVNGKV KGRIFVGSSQ VPIVFENTDL HSYVVMNHGR SYTAISTIPE TVGYSLLPLA PVGGIIGWMF AVEQDGFKNG FSITGGEFTR QAEVTFVGHP GNLVIKQRFS GIDEHGHLTI DTELEGRVPQ IPFGSSVHIE PYTELYHYST SVITSSSTRE YTVTEPERDG ASPSRIYTYQ WRQTITFQEC VHDDSRPALP STQQLSVDSV FVLYNQEEKI LRYALSNSIG PVREGSPDAL QNPCYIGTHG CDTNAACRPG PRTQFTCECS IGFRGDGRTC YDIDECSEQP SVCGSHTICN NHPGTFRCEC VEGYQFSDEG TCVAVVDQRP INYCETGLHN CDIPQRAQCI YTGGSSYTCS CLPGFSGDGQ ACQDVDECQP SRCHPDAFCY NTPGSFTCQC KPGYQGDGFR CVPGEVEKTR CQHEREHILG AAGATDPQRP IPPGLFVPEC DAHGHYAPTQ CHGSTGYCWC VDRDGREVEG TRTRPGMTPP CLSTVAPPIH QGPAVPTAVI PLPPGTHLLF AQTGKIERLP LEGNTMRKTE AKAFLHVPAK VIIGLAFDCV DKMVYWTDIT EPSIGRASLH GGEPTTIIRQ DLGSPEGIAV DHLGRNIFWT DSNLDRIEVA KLDGTQRRVL FETDLVNPRG IVTDSVRGNL YWTDWNRDNP KIETSYMDGT NRRILVQDDL GLPNGLTFDA FSSQLCWVDA GTNRAECLNP SQPSRRKALE GLQYPFAVTS YGKNLYFTDW KMNSVVALDL AISKETDAFQ PHKQTRLYGI TTALSQCPQG HNYCSVNNGG CTHLCLATPG SRTCRCPDNT LGVDCIEQK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.