missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Nidogen Recombinant Protein, PeproTech®
Description
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
Specifications
Specifications
| Accession Number | P14543 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 4811 |
| Molecular Weight (g/mol) | 133.5 kDa |
| Name | Human Nidogen |
| Quantity | 5 μg |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?