missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NHS Partial ORF (NP_938011.1, 1532 a.a. - 1629 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004810-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16181925

  • 335.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein containing four conserved nuclear localization signals. The encoded protein may function during the development of the eyes, teeth, and brain. Mutations in this gene have been shown to cause Nance-Horan syndrome. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: TAESPQSTDDAHQGSQGAEALSPLSPCSPRVNAEGFSSKSFATSASARVGRSRAPPAASSSRYSVRCRLYNTPMQAISEGETENSDGSPHDDRSSQSS
Specifications

Specifications

NP_938011.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TAESPQSTDDAHQGSQGAEALSPLSPCSPRVNAEGFSSKSFATSASARVGRSRAPPAASSSRYSVRCRLYNTPMQAISEGETENSDGSPHDDRSSQSS
RUO
NHS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4810
NHS (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781F2016/DKFZp781L0254/SCML1
NHS
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions